missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06855AF594
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifikationer
| Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B | |
| Partial recombinant protein made to an internal portion of the human JMJD3 protein (amino acids 1520-1682) [UniProt O15054] | |
| 0.1 mL | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 23135 | |
| Store at 4°C in the dark. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering