Läs mer
Lysinoalanine Dihydrochloride, TRC
Produktkod. 30468671
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30468671 100 mg
1 options
Produktkod. 30468671 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL503198100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lysinoalanine dihydrochloride
CAS 4418-81-9
Molekylformel C9 H19 N3 O4 . 2 Cl H
Kvantitet 100 mg
Synonym N6-(2-Amino-2-carboxyethyl)-L-lysin Dihydrochloride
InChI formel InChI=1S/C9H19N3O4.2ClH/c10-6(8(13)14)3-1-2-4-12-5-7(11)9(15)16;;/h6-7,12H,1-5,10-11H2,(H,13,14)(H,15,16);2*1H/t6-,7?;;/m0../s1
LEDER Cl.Cl.N[C@@H](CCCCNCC(N)C(=O)O)C(=O)O
IUPAC-namn (2S)-2-amino-6-[(2-amino-2-carboxyethyl)amino]hexanoic acid;dihydrochloride
Molekylvikt (g/mol) 306.19
Formel vikt 305.0909