Läs mer
Lysinoalanine Hydrochloride Salt, TRC
Produktkod. 30136267
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136267 50 mg
30007916 10 mg
30006861 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136267 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL48865050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93113-28-1
Molekylformel C9H19N3O4.xHCl
Molekylvikt (g/mol) 269.73
Synonym N6-(2-Amino-2-carboxyethyl)-L-lysine Hydrochloride Salt;
IUPAC-namn N6-(2-amino-2-carboxyethyl)-L-lysine Hydrochloride Salt;
LEDER N[C@@](C(O)=O)([H])CCCCNCC(N)([H])C(O)=O.Cl
Specifikationer
Kemiskt namn eller material Lysinoalanine Hydrochloride Salt
Anteckningar om renhetsgrad HPLC
InChI formel N[C@@](C(O)=O)([H])CCCCNCC(N)([H])C(O)=O.[xHCl]
Kvantitet 50 mg
Formel vikt 269.11
Procent renhet >95%