Läs mer
MedChemExpress Lysipressin
Produktkod. 16419318
Encompass_Preferred
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
16419318 10 mg
1 options
Produktkod. 16419318 Leverantör MedChemExpress Leverantörsnummer HYP0004
MedChemExpress Lysipressin is Antidiuretic hormone that have been found in pigs and some marsupial families. Induces contraction of the rabbit urinary bladder smooth muscle, activate adenylate-cyclase.
Specifikationer
CAS 50-57-7
Innehåll och lagring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molekylvikt (g/mol) 1056.22 g/mol
Produkttyp Natural Products
Kvantitet 10 mg
Frakt skick Room temperature
Löslighet H2O : 100 mg/mL (94.68 mM∣Need ultrasonic)
Synonym Lysine vasopressin∣ [Lys8]-Vasopressin