missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYZL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11922-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Lysozymes.Specifications
LYZL6 | |
Polyclonal | |
Unconjugated | |
LYZL6 | |
1700023H08Rik, LYC1, lysozyme like 6, Lysozyme like protein 6, LYZL6, PRO1485, TKAL754 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287751, 57151, 69444 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75951, Q9DA11 | |
Lyzl6 | |
LYZL6 Fusion Protein Ag2569 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |