missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565086
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VLKYTAQNMEL QNKVQLLEEQ NLSLLDQLRK LQAMVIEISN KTS Highest antigen sequence identity to the following orthologs - mouse 93%, rat 91%.
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds to the cAMP-responsive element, an octameric palindrome. The protein interacts with host cell factor C1, which also associates with the herpes simplex virus (HSV) protein VP16 that induces transcription of HSV immediate-early genes. This protein and VP16 both bind to the same site on host cell factor C1. It is thought that the interaction between this protein and host cell factor C1 plays a role in the establishment of latency during HSV infection. An additional transcript variant has been identified, but its biological validity has not been determined.Specifications
LZIP | |
Polyclonal | |
Unconjugated | |
CREB3 | |
AU044960; AW538053; basic leucine zipper protein; C80076; cAMP responsive element binding protein 3; cAMP-responsive element-binding protein 3; CREB3; CREB-3; cyclic AMP response element (CRE)-binding protein/activating transcription factor 1; cyclic AMP-responsive element-binding protein 3; leucin zipper proitein; Leucine zipper protein; Luman; LZIP; LZIP-1; LZIP-2; N-terminal Luman; Processed cyclic AMP-responsive element-binding protein 3; sLZIP; small leucine zipper protein; transcription factor LZIP; transcription factor LZIP-alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43889 | |
CREB3 | |
Recombinant Human LZIP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only