Learn More
LZTS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15677-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 7p.Specifications
LZTS2 | |
Polyclonal | |
Unconjugated | |
LZTS2 | |
hLZTS2, KIAA1813, LAPSER1, LZTS2, Protein LAPSER1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
226154, 365468, 84445 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3LUD4, Q91YU6, Q9BRK4 | |
Lzts2 | |
LZTS2 Fusion Protein Ag8145 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |