Läs mer
Tocris Bioscience™ M 084 hydrochloride
Beskrivning
M 084 hydrochloride is a TRPC4/5 channel blocker (IC50 values are 3.7-10.3 and 8.2 μM, respectively). Also weakly blocks TRPC3 channels. Exhibits rapid antidepressant and anxiolytic effects in vivo.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Butyl-1H-benzimidazol-2-amine hydrochloride |
| CAS | 1992047-63-8 |
| Kvantitet | 10 mg |
| Mål | TRPC Channel Blockers |
| Molekylformel | C11H15N3.HCl |
| Formel vikt | 225.72 |
| Renhet | >99% |