Läs mer
m-Aminophenyl Tosylate, TRC
Produktkod. 30312312
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30312312 1 g
1 options
Produktkod. 30312312 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6270701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material m-Aminophenyl Tosylate
CAS 3865-15-4
Molekylformel C13H13NO3S
Kvantitet 1 g
Synonym 3-Aminophenol 4-Methylbenzenesulfonate (Ester), m-Aminophenol p-Toluenesulfonate, m-Aminophenol p-Toluenesulfonate (Ester)
InChI formel InChI=1S/C13H13NO3S/c1-10-5-7-13(8-6-10)18(15,16)17-12-4-2-3-11(14)9-12/h2-9H,14H2,1H3
LEDER CCOC(=O)C1N=C(c2c(F)cccc2)c3c(ccc(Cl)c3)NC1=O
IUPAC-namn (3-aminophenyl) 4-methylbenzenesulfonate
Molekylvikt (g/mol) 263.31