Läs mer
m-Chloro Hippuric Acid, TRC
Produktkod. 30366909
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366909 10 g
30353182 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366909 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36655510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57728-59-3
Molekylformel C9H8ClNO3
Molekylvikt (g/mol) 213.62
Synonym (3-Chlorobenzoylamino)acetic Acid, N-(3-Chlorobenzoyl)glycine, NSC 201882, m-Chlorohippuric Acid, N-(3-Chlorobenzoyl)glycine
IUPAC-namn 2-[(3-chlorobenzoyl)amino]acetic acid
LEDER OC(=O)CNC(=O)c1cccc(Cl)c1
Specifikationer
Kemiskt namn eller material m-Chloro Hippuric Acid
Kvantitet 10 g
InChI formel InChI=1S/C9H8ClNO3/c10-7-3-1-2-6(4-7)9(14)11-5-8(12)13/h1-4H,5H2,(H,11,14)(H,12,13)
Formel vikt 213.0193