Läs mer
m-Cumenyl Phosphate, TRC
Produktkod. 30364522
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364522 50 mg
30368445 500 mg
30350233 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364522 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC83465050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72668-27-0
Molekylformel C27 H33 O4 P
Molekylvikt (g/mol) 452.52
Synonym m-Cumenyl phosphate (7CI), Tris(3-isopropylphenyl) phosphate, Tris(m-isopropylphenyl) phosphate, Phenol, 3-(1-methylethyl)-, phosphate (3:1) (9CI), PIP (3:1)
IUPAC-namn tris(3-isopropylphenyl) phosphate
LEDER CC(C)c1cccc(OP(=O)(Oc2cccc(c2)C(C)C)Oc3cccc(c3)C(C)C)c1
Specifikationer
Kemiskt namn eller material Tris[3-(2-methylethyl)phenyl] Phosphate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C27H33O4P/c1-19(2)22-10-7-13-25(16-22)29-32(28,30-26-14-8-11-23(17-26)20(3)4)31-27-15-9-12-24(18-27)21(5)6/h7-21H,1-6H3
Formel vikt 452.2116
Procent renhet >95%