missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Ethyltoluene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -97°C to 95°C |
Density | 0.8600g/mL |
Boiling Point | 158.0°C to 159.0°C |
Flash Point | 38°C |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | C2H5C6H4CH3 |
Refractive Index | 1.4956 |
Quantity | 25 g |
Beilstein | 05,396 |
Specific Gravity | 0.86 |
Show More |
Safety and Handling
Product Identifier
- m-Ethyltoluene, 98%
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'