Läs mer
m-Fluoro Prasugrel Hydrochloride, TRC
Produktkod. 30009638
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009638 10 mg
30008189 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009638 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF59587010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391052-75-7
Molekylformel C20 H20 F N O3 S . Cl H
Molekylvikt (g/mol) 409.9
Synonym Ethanone, 2-[2-(acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-cyclopropyl-2-(3-fluorophenyl)-, hydrochloride (1:1), 2-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-cyclopropyl-2-(3-fluorophenyl)ethanone hydrochloride (1:1), m-Fluoro Prasugrel hydrochloride, m-Fluoroprasugrel hydrochloride, 5-[(1RS)-2-Cyclopropyl-1-(3-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate hydrochloride, 5-[2-Cyclopropyl-1-(3-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate hydrochloride
IUPAC-namn [5-[2-cyclopropyl-1-(3-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate;hydrochloride
LEDER Cl.CC(=O)Oc1cc2CN(CCc2s1)C(C(=O)C3CC3)c4cccc(F)c4
Specifikationer
Kemiskt namn eller material m-Fluoro Prasugrel Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C20H20FNO3S.ClH/c1-12(23)25-18-10-15-11-22(8-7-17(15)26-18)19(20(24)13-5-6-13)14-3-2-4-16(21)9-14;/h2-4,9-10,13,19H,5-8,11H2,1H3;1H
Formel vikt 409.0915