Läs mer
m,m-Quaterphenyl, TRC
Produktkod. 30426664
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426664 500 mg
30419508 50 mg
30416417 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ116570500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1166-18-3
Molekylformel C24H18
Molekylvikt (g/mol) 306.4
Synonym m-Tetraphenyl, 1-Phenyl-3-(3-phenylphenyl)benzene, 3,3'-Diphenylbiphenyl, NSC 90718
IUPAC-namn 1-phenyl-3-(3-phenylphenyl)benzene
LEDER c1ccc(cc1)c2cccc(c2)c3cccc(c3)c4ccccc4
Specifikationer
Kemiskt namn eller material m,m-Quaterphenyl
Kvantitet 500 mg
InChI formel InChI=1S/C24H18/c1-3-9-19(10-4-1)21-13-7-15-23(17-21)24-16-8-14-22(18-24)20-11-5-2-6-12-20/h1-18H
Formel vikt 306.1409