missing translation for 'onlineSavingsMsg'
Läs mer

m-Opiazone, TRC

Produktkod. 30411312
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411312 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30411312 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO669500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material m-Opiazone
CAS 4821-88-9
Molekylformel C10H10N2O3
Kvantitet 100 mg
Synonym 6,7-Dimethoxyphthalazin-1(2H)-one
InChI formel InChI=1S/C10H10N2O3/c1-14-8-3-6-5-11-12-10(13)7(6)4-9(8)15-2/h3-5H,1-2H3,(H,12,13)
LEDER COc1cc2C=NNC(=O)c2cc1OC
IUPAC-namn 6,7-dimethoxy-2H-phthalazin-1-one
Molekylvikt (g/mol) 206.2
Formel vikt 206.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.