Learn More
Brady™ M Series MetaLabel™ Solvent Resistant Metallized Polyester Labels, Black on Light Gray
Metallized Polyester (B-428) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
Brand:  Brady™ 143278
Additional Details : Weight : 0.11340kg
 View more versions of this product
Product Code. 11992792
  • 733.00 SEK / 1 box
Estimated Shipment: 23-08-2021
Add to basket
Description
Description
  • Matte, light gray finish gives labels a rating and serial plate appearance
  • Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • UL approved
  • CSA accepted
  • All-in-one cartridge includes label supply and R4300 Series ribbon
Specifications
Specifications
Label
Permanent Acrylic
ASTM D 1000; ASTM D 2979; ASTM G 155; CSA; Halogen Free (DIN VDE 0472 Part 815); UL Recognized
1.5 in.
1
1.5 x 0.5 in. (W x H)
Metallized Polyester
1.5 x 0.5 in.
0.5 in.
69.85 mm
Not Autoclavable
Light Gray
Label
BMP™51 Label Maker, BMP™53 Label Maker
Rectangle
No Text
360 Labels/Pk.
Cartridge, Ribbon
Thermal Transfer