missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Description
Reactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents.
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents.
Specifications
Boiling Point | 252°C to 253°C |
Flash Point | 107°C (224°F) |
Odor | Acrid |
Quantity | 1 g |
UN Number | UN3265 |
Beilstein | 2087865 |
Formula Weight | 190.65 |
Percent Purity | 98% |
Chemical Name or Material | m-Toluenesulfonyl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'