Läs mer
m-Tyramine Hydrochloride, TRC
Produktkod. 30453918
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30453918 100 mg
1 options
Produktkod. 30453918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT898505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material m-Tyramine Hydrochloride
CAS 3458-98-8
Molekylformel C8H11NO .HCl
Kvantitet 100 mg
Synonym 3-Hydroxyphenethylamine Hydrochloride, 3-(2-Aminoethyl)phenol Hydrochloride, 2-(3-Hydroxyphenyl)ethylamine Hydrochloride, 2-(3-Hydroxyphenyl)ethanamine Hydrochloride, m-(2-Aminoethyl)-phenol Hydrochloride, 3-(2-Aminoethyl)-phenol Hydrochloride
InChI formel InChI=1S/C8H11NO.ClH/c9-5-4-7-2-1-3-8(10)6-7;/h1-3,6,10H,4-5,9H2;1H
LEDER Cl.NCCc1cccc(O)c1
IUPAC-namn 3-(2-aminoethyl)phenol;hydrochloride
Molekylvikt (g/mol) 173.64
Formel vikt 173.0607