Läs mer
m-Vinylaniline (contains KOH as inhibitor), TRC
Produktkod. 30451760
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30451760 50 mg
1 options
Produktkod. 30451760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV36872550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material m-Vinylaniline
CAS 15411-43-5
Molekylformel C8H9N
Kvantitet 50 mg
Synonym 3-Ethenylbenzenamine, 3-Aminostyrene, 3-Vinylaniline, m-Aminostyrene
InChI formel InChI=1S/C8H9N/c1-2-7-4-3-5-8(9)6-7/h2-6H,1,9H2
LEDER C=CC1=CC(=CC=C1)N
IUPAC-namn 3-ethenylaniline
Molekylvikt (g/mol) 119.16
Formel vikt 119.0735