Läs mer
Brady™ M611 Label Printer - EU with Brady Workstation LAB Suite
Produktkod. 17137001
:
Resolution: 300 dpi
Produktkod. 17137001
Brand: Brady™ 306858
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A smartphone operated printer for cables and components, laboratory samples and complex labeling, electrical and datacom identification using the Brady Workstation Mobile Express Labels app.
  • The BradyPrinter M611 Mobile Printer is a smartphone operated printer for cables and components, laboratory samples and complex labeling, electrical and datacom identification using the Brady Workstation Mobile Express Labels app.
  • You can easily create 1D and 2D codes and barcodes with a smartphone, serialise, use an extensive image library, include text, a time stamp and even data from the cloud in a complex label that can easily be designed in just a couple of steps.
  • The Express app offers the most comprehensive label design capabilities available for any smartphone and includes design wizards to quickly create specific identification labels.
  • Join the evolution of smarter identification.
  • The printer comes with the Brady Workstation Laboratory Identification Software suite enabling you to immediately start designing professional labels.
  • This suite combines frequently used software apps to create laboratory identification labels.
Specifikationer
Visningstyp Backlit Color LCD Touchscreen
Tryckteknik Thermal Transfer
Bredd (metrisk) 113.00 mm
Höjd (metrisk) 115.00 mm
Färgkapacitet Monochrome
Inkluderar USB cable, a quick start guide, an AC powercord, a USB stick holding product info and Windows driver, a strap, a Brady Workstation Laboratory Identification Suite, a roll of PTL-97-488 and an M61-R4310 ribbon
Utskriftshastighet 33.80 mm/s
Upplösning 300 dpi
Strömförsörjningsdetaljer AC Adaptor, Rechargeable Battery