Läs mer
(-)-Maackiain, TRC
Produktkod. 30479245
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30479245 5 mg
1 options
Produktkod. 30479245 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2143905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-Maackiain
CAS 2035-15-6
Molekylformel C16 H12 O5
Kvantitet 5 mg
Synonym 6H-[1,3]Dioxolo[5,6]benzofuro[3,2-c][1]benzopyran-3-ol, 6a,12a-dihydro-, (6aR,12aR)-, 6H-[1,3]Dioxolo[5,6]benzofuro[3,2-c][1]benzopyran-3-ol, 6a,12a-dihydro-, (6aR-cis)-, 6H-[1,3]Dioxolo[5,6]benzofuro[3,2-c][1]benzopyran-3-ol, 6aα,12aα-dihydro-, (-)- (8CI), Maackiain (7CI), (-)-(6aR,12aR)-maackiain, (-)-Maackiain, (6aR,12aR)-6a,12a-Dihydro-6H-[1,3]dioxolo[5,6]benzofuro[3,2-c][1]benzopyran-3-ol, Inermin, Inermine, Maackiaine, Trifolirhizin aglycone, l-Maackiain
InChI formel InChI=1S/C16H12O5/c17-8-1-2-9-12(3-8)18-6-11-10-4-14-15(20-7-19-14)5-13(10)21-16(9)11/h1-5,11,16-17H,6-7H2/t11-,16-/m0/s1
LEDER Oc1ccc2[C@@H]3Oc4cc5OCOc5cc4[C@@H]3COc2c1
Molekylvikt (g/mol) 284.26
Formel vikt 284.0685