Läs mer
Mabuterol Hydrochloride, TRC
Produktkod. 30480905
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30480905 1 mg
1 options
Produktkod. 30480905 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1040101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mabuterol hydrochloride
CAS 54240-36-7
Molekylformel C13 H18 Cl F3 N2 O . Cl H
Kvantitet 1 mg
Synonym Benzenemethanol, 4-amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)-, monohydrochloride (9CI), Broncholin, KF 868, KF 868 (pharmaceutical), dl-Mabuterol hydrochloride, Benzenemethanol, 4-amino-3-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-5-(trifluoromethyl)-, hydrochloride (1:1)
InChI formel InChI=1S/C13H18ClF3N2O.ClH/c1-12(2,3)19-6-10(20)7-4-8(13(15,16)17)11(18)9(14)5-7;/h4-5,10,19-20H,6,18H2,1-3H3;1H
LEDER Cl.CC(C)(C)NCC(O)c1cc(Cl)c(N)c(c1)C(F)(F)F
IUPAC-namn 1-[4-amino-3-chloro-5-(trifluoromethyl)phenyl]-2-(tert-butylamino)ethanol;hydrochloride
Molekylvikt (g/mol) 347.2
Formel vikt 346.0827