Beskrivning
MafA Polyclonal antibody specifically detects MafA in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Gel Super Shift Assays,ChIP assay (ChIP),Immunocytochemistry/ Immunofluorescence
Specifikationer
Specifikationer
| Antigen | MafA |
| Användningsområden | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Supershift, ChIP Assay, Immunocytochemistry/Immunofluorescence |
| Klassificering | Polyclonal |
| Konjugera | PE |
| Formulering | PBS |
| Gene Alias | hMafA, Pancreatic beta-cell-specific transcriptional activator, RIPE3B1, transcription factor MafA, Transcription factor RIPE3b1, V-maf musculoaponeurotic fibrosarcoma oncogene homolog A, v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian) |
| Värd art | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 300 and the C-terminus (residue 359) of mouse v-maf musculoaponeurotic fibrosarcoma oncogene homolog A using the numbering given in entry NP_919331.1 (GeneID 378435). |
| Reningsmetod | Immunogen affinity purified |
| Kvantitet | 0.1 mL |
| Visa mer |