Läs mer
MAFB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20189-1-AP-150UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.Specifikationer
| MAFB | |
| Polyclonal | |
| Unconjugated | |
| MAFB | |
| KRML, Maf B, MAFB, Transcription factor MafB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16658, 54264, 9935 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P54841, P54842, Q9Y5Q3 | |
| Mafb | |
| MAFB Fusion Protein Ag14102 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |