Learn More
Mafosfamide Sodium Salt, TRC
Product Code. 30007978
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007978 10 mg 10mg
30012838 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30007978 Supplier Toronto Research Chemicals Supplier No. TRCM11030010MG
 Restricted Item
Beräknad leverans: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84211-05-2
Molecular Formula C9H18Cl2N2NaO5PS2
Molecular Weight (g/mol) 423.24
Synonym rel-2-[[(2R,4R)-2-[Bis(2-chloroethyl)amino]tetrahydro-2-oxido-2H-1,3,2-oxazaphosphorin-4-yl]thio]ethanesulfonic Acid Sodium, Mafosfamid Sodium, Z 7557, cis-Mafosfamide Sodium;
IUPAC Name sodium 2-(((2R,4R)-2-(bis(2-chloroethyl)amino)-2-oxido-1,3,2-oxazaphosphinan-4-yl)thio)ethane-1-sulfonate
SMILES ClCCN(P1(N[C@@H](CCO1)SCCS(=O)([O-])=O)=O)CCCl.[Na+]
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C9H19Cl2N2O5PS2.Na/c10-2-4-13(5-3-11)19(14)12-9(1-6-18-19)20-7-8-21(15,16)17;/h9H,1-8H2,(H,12,14)(H,15,16,17);/q;+1/p-1/t9-,19-;/m1./s1
Formula Weight 421.97
Chemical Name or Material Mafosfamide Sodium Salt (Mixture of Diastereomers)