missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MAGEB4 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850684
|
Proteintech
12786-2-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840684
|
Proteintech
12786-2-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEB genes are clustered on chromosome Xp22-p21. This gene sequence ends in the first intron of MAGEB1, another family member. This gene is expressed in testis.Specifications
MAGEB4 | |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
O15481 | |
4115 | |
MAGEB4 Fusion Protein Ag3502 | |
Primary | |
-20°C | |
MAGEB4 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CT3.6, MAGE B4 antigen, MAGEB4, melanoma antigen family B, 4, Melanoma associated antigen B4 | |
MAGEB4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |