Läs mer
Magnesium 2-Aminoacetate, TRC
Produktkod. 30003776
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003776 100 mg
30010170 10 mg
30010266 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003776 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB475003100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14783-68-7
Molekylformel C4H8MgN2O4
Molekylvikt (g/mol) 172.42
Synonym Magnesium Glycinate
IUPAC-namn magnesium;2-aminoacetate
LEDER [Mg+2].NCC([O-])=O.NCC([O-])=O
Specifikationer
Kemiskt namn eller material Magnesium 2-Aminoacetate
Kvantitet 100 mg
InChI formel InChI=1S/2C2H5NO2.Mg/c2*3-1-2(4)5;/h2*1,3H2,(H,4,5);/q;;+2/p-2
Formel vikt 172.0334