Learn More
Magnesium Acetate Tetrahydrate, Supelco™
Product Code. 15867400
Change view
Click to view available options
Quantity:
250 g
1 kg
50 kg
Unit Size:
1kg
250g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867400 50 kg 50kg
15847400 250 g 250g
15857400 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15867400 Supplier Supelco Supplier No. 1058199050
 Restricted Item
Beräknad leverans: 19-11-2026
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16674-78-5
Specifications
Melting Point 80°C
Quantity 50 kg
Molecular Formula (CH3COO)2Mg · 4 H2O
MDL Number MFCD00149214
Synonym Magnesium acetate tetrahydrate, Magnesium acetate tetrahydrate
Molecular Weight (g/mol) 214.45
Grade Research
Density 1.45 g/cm3
Chemical Name or Material Magnesium Acetate Tetrahydrate