missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10518060
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
This item is not returnable. View return policy
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
Melting Point 117°C
Color White
pH 5 to 6.5
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 203.31g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'