missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium fluoride, 99% (metals basis excluding Ca & Na), Ca+Na <1%, Thermo Scientific Chemicals
633.53 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1261°C |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99%, Ca+Na <1% (Metals basis excluding Ca and Na) |
Solubility Information | Sparingly soluble in water. Slightly soluble in dilute HNO3Insoluble in ethanol. |
Formula Weight | 62.3 |
Odor | Odorless |
Refractive Index | 1.35-1.38 |
Density | 3.148 g/cm3 at 20°C |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'