missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Nitrate Hexahydrate, Certified AR for Analysis, 99.0-101.0%, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Specifications
Boiling Point | 330°C |
Melting Point | 89°C |
Color | White |
pH | 5.0 to 8.2 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 256.4g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'