missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Magnesium oxide is used as a laboratory reagent, and a good reference white color for colorimetry. Magneisum oxide microspheres is used for enrichment method of polycyclic aromatic hydrocarbons. It is also used in groundwater studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 3600°C |
Melting Point | 2852°C |
Physical Form | Fused Lump |
Quantity | 25 g |
Merck Index | 14,5677 |
Solubility Information | Soluble in HCl and dilute acids. Insoluble in water and ethanol. |
Formula Weight | 40.3 |
Percent Purity | 99.95% |
Odor | Odorless |
Refractive Index | 1.736 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'