missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | >310°C |
Linear Formula | Mg(CF3SO3)2 |
Quantity | 5 g |
UN Number | UN1759 |
Beilstein | 705578 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 322.43 |
Percent Purity | 98% |
Chemical Name or Material | Magnesium trifluoromethanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'