Läs mer
Magnesium Valproate, TRC
Produktkod. 30135719
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135719 10 g
17960775 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135719 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM11035010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 62959-43-7
Molekylformel C16 H30 Mg O4
Molekylvikt (g/mol) 310.71
Synonym Dipromal, Magnesium Dipropylacetate, 2-Propylpentanoic Acid Magnesium
IUPAC-namn bis(2-propylpentanoyloxy)magnesium
LEDER CCCC(CCC)C(=O)O[Mg]OC(=O)C(CCC)CCC
Specifikationer
Kemiskt namn eller material Magnesium Valproate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/2C8H16O2.Mg/c2*1-3-5-7(6-4-2)8(9)10;/h2*7H,3-6H2,1-2H3,(H,9,10);/q;;+2/p-2
Formel vikt 310.1995
Procent renhet >95%