Learn More
Invitrogen™ MagniSort™ Streptavidin Negative Selection Beads
Designed for the magnetic separation of cells by negative selection. Undesired cells are bound by a cocktail of biotinylated antibodies and then streptavidin coated magnetic beads.
Brand: Invitrogen™ MSNB-6002-71
609.36 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Description
- Clone and concentration of biotinylated antibody for undesired cell labeling, as well as concentration of MagniSort™ Streptavidin Negative Selection Beads must be determined empirically
- Please refer to the protocol for details on how to optimize both the antibody or antibody cocktail and MagniSort™ Streptavidin Negative Selection Beads.
- For depletion of cells using a single antibody, we recoomend the MagniSort™ Streptavidin Positive Selection Beads (cat. MSPB-6003) and protocol
Specifications
25 Tests | |
Streptavidin | |
Biotin |
Liquid | |
Streptavidin Conjugate (Fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.