Läs mer
Magnoflorine Hydroxide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Magnoflorine Hydroxide |
| CAS | 476-63-1 |
| Molekylformel | C20H25NO5 |
| Kvantitet | 5 mg |
| Synonym | (6aS)-5,6,6a,7-Tetrahydro-1,11-dihydroxy-2,10-dimethoxy-6,6-dimethyl-4H-dibenzo[de,g]quinolinium Hydroxide, (+)-Magnoflorine Hydroxide, Escholin Hydroxide, Escholine Hydroxide, Magnoflorine Hydroxide, Thalictrin Hydroxide, Thalictrine Hydroxide, α-Magnoflorine Hydroxide |
| LEDER | [OH-].COc1ccc2C[C@H]3c4c(CC[N+]3(C)C)cc(OC)c(O)c4c2c1O |
| Molekylvikt (g/mol) | 359.42 |