missing translation for 'onlineSavingsMsg'
Learn More

MAGOH Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1355.00 SEK - 3665.00 SEK

Specifications

Antigen MAGOH
Concentration 0.13 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16840014
View Documents Promotion Details
Proteintech
12347-1-AP-150UL
150 μL
3665.00 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 06-12-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16850014
View Documents Promotion Details
Proteintech
12347-1-AP-20UL
20 μL
1355.00 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 06-12-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Drosophila that have mutations in their mago nashi gene produce progeny with defects in germplasm assembly and germline development. This gene encodes the mammalian mago nashi homolog. In mammals, mRNA expression is not limited to the germ plasm, but is expressed ubiquitously in adult tissues and can be induced by serum stimulation of quiescent fibroblasts.
Specifications

Specifications

MAGOH
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Human
P61326
4116
MAGOH Fusion Protein Ag3004
Primary
-20°C
MAGOH
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
MAGOH, MAGOHA, Protein mago nashi homolog
MAGOH
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications