Learn More
Cytiva Mains Cable
Mains cable accessory for AKTA start preparative chromatography system
200.66 SEK - 904.56 SEK
Specifications
For Use With (Equipment) | KTA Start Preparative Chromatography System for Laboratory Scale Protein Purification |
---|---|
Type | Mains Cable |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15219794
|
Cytiva
19244701 |
120 V |
868.63 SEK
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
15229794
|
Cytiva
19244801 |
220 V |
904.56 SEK
Each |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
16525850
|
Cytiva
18-1100-12 |
CH Version |
200.66 SEK
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Accessory for the AKTA start preparative chromatography system forlaboratory scale protein purification from Cytiva.Specifications
KTA Start Preparative Chromatography System for Laboratory Scale Protein Purification | |
Mains Cable |