Läs mer
MAK683, MedChemExpress
Produktkod. 16492886
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16492886 25 mg
16466607 5 mg
16460247 50 mg
16413888 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16492886 Leverantör Medchem Express Leverantörsnummer HY103663
MedChemExpress MAK683 is an embryonic ectoderm development (EED) inhibitor extracted from patent US20160176882 A1, compound example 2. MAK683 exhibits IC50s of 59, 89, 26 nM in EED Alphascreen binding, LC-MS and ELISA assay.
MAK683 is an embryonic ectoderm development (EED) inhibitor extracted from patent US20160176882 A1, compound example 2. MAK683 exhibits IC50s of 59, 89, 26 nM in EED Alphascreen binding, LC-MS and ELISA assay.
Specifikationer
Kemiskt namn eller material MAK683
CAS 1951408-58-4
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C20H17FN6O
Kvantitet 25 mg
Löslighetsinformation DMSO : 52 mg/mL (138.15 mM; Need ultrasonic)
LEDER CC1=NC=CC=C1C2=CN=C(NCC3=C4CCOC4=CC=C3F)N5C2=NN=C5
Molekylvikt (g/mol) 376.39
Formel vikt 376.39
Procent renhet 99.27%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death