Läs mer
MAL-PEG2-NHS Ester, TRC
Produktkod. 30010123
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010123 25 mg -
30007332 2.5 mg 2.5mg
30009709 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010123 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM10000525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1433997-01-3
Molekylformel C15 H18 N2 O8
Molekylvikt (g/mol) 354.312
Synonym 2,5-Dioxo-1-pyrrolidinyl 3-[2-[2-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)ethoxy]ethoxy]propanoate, 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(2,5-Dioxo-2H-pyrrol-1(5H)-yl)ethoxy)ethoxy)propanoate, 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)ethoxy)propanoate
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 3-[2-[2-(2,5-dioxopyrrol-1-yl)ethoxy]ethoxy]propanoate
LEDER O=C(CCOCCOCCN1C(=O)C=CC1=O)ON2C(=O)CCC2=O
Specifikationer
Kemiskt namn eller material MAL-PEG2-NHS Ester
Kvantitet 25 mg
InChI formel InChI=1S/C15H18N2O8/c18-11-1-2-12(19)16(11)6-8-24-10-9-23-7-5-15(22)25-17-13(20)3-4-14(17)21/h1-2H,3-10H2
Formel vikt 354.106