Läs mer
Malabaricone B, TRC
Produktkod. 30466804
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30466804 1 mg
30471039 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30466804 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2138351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Malabaricone B
CAS 63335-24-0
Molekylformel C21 H26 O4
Kvantitet 1 mg
Synonym 1-(2,6-Dihydroxyphenyl)-9-(4-hydroxyphenyl)-1-nonanone, 1-(2,6-Dihydroxyphenyl)-9-(4-hydroxyphenyl)nonan-1-one, NSC 287967, NSC 630196
InChI formel InChI=1S/C21H26O4/c22-17-14-12-16(13-15-17)8-5-3-1-2-4-6-9-18(23)21-19(24)10-7-11-20(21)25/h7,10-15,22,24-25H,1-6,8-9H2
LEDER Oc1ccc(CCCCCCCCC(=O)c2c(O)cccc2O)cc1
IUPAC-namn 1-(2,6-dihydroxyphenyl)-9-(4-hydroxyphenyl)nonan-1-one
Molekylvikt (g/mol) 342.43
Formel vikt 342.1831