Learn More
Abnova™ MALAT1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
19880.00 SEK - 32740.00 SEK
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | metastasis associated lung adenocarcinoma transcript 1 (non-protein coding) |
Gene Alias | HCN,MALAT-1,NCRNA00047,NEAT2,PRO2853 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063515
|
Abnova™
FS0065.100UL |
100 μL |
19880.00 SEK
100µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073515
|
Abnova™
FS0065.200UL |
200 μL |
32740.00 SEK
200µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
HCN,MALAT-1,NCRNA00047,NEAT2,PRO2853 | |
378938 | |
Liquid |
Tube | |
metastasis associated lung adenocarcinoma transcript 1 (non-protein coding) | |
MALAT1 | |
DNA/RNA |