Läs mer
Malathion beta-Monoacid, TRC
Produktkod. 30467027
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30467027 100 mg
1 options
Produktkod. 30467027 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM111020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Malathion beta-Monoacid
CAS 1642-51-9
Molekylformel C8 H15 O6 P S2
Kvantitet 100 mg
Synonym 2-[(Dimethoxyphosphinothioyl)thio]butanedioic Acid 1-Ethyl Ester, Malathion β-Monocarboxylic Acid, O,O-Dimethyl S-(1-Carbethoxy-2-carboxy)ethyl Phosphorodithioate, S-[2-Carboxy-1-(ethoxycarbonyl)ethyl]O,O-di-methyl Phosphorodithioate, Malathion Monoacid
InChI formel InChI=1S/C8H15O6PS2/c1-4-14-8(11)6(5-7(9)10)17-15(16,12-2)13-3/h6H,4-5H2,1-3H3,(H,9,10)
LEDER CCOC(=O)C(CC(=O)O)SP(=S)(OC)OC
IUPAC-namn 3-dimethoxyphosphinothioylsulfanyl-4-ethoxy-4-oxo-butanoic acid
Molekylvikt (g/mol) 302.3
Formel vikt 302.0048