missing translation for 'onlineSavingsMsg'
Learn More

Malonic acid disodium salt, 99%, Thermo Scientific Chemicals

Catalog Number p-7059553
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
141-95-7
C3H2Na2O4
148.025
MFCD00002708
PRWXGRGLHYDWPS-UHFFFAOYSA-L
sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
8865
CHEBI:62983
disodium;propanedioate
C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
141-95-7
C3H2Na2O4
148.025
MFCD00002708
PRWXGRGLHYDWPS-UHFFFAOYSA-L
sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
8865
CHEBI:62983
disodium;propanedioate
C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Malonic acid disodium salt is used in organic Synthesis, pharmaceuticals, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Solubility Information Soluble in water (148g/L at 20°C).
Formula Weight 148.03
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Malonic acid disodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'