Läs mer
Malvidin Chloride, TRC
Produktkod. 30012579
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012579 5 mg
30008664 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012579 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1615305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 643-84-5
Molekylformel C17 H15 O7 . Cl
Molekylvikt (g/mol) 366.75
Synonym 1-Benzopyrylium, 3,5,7-trihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-, chloride (1:1), 1-Benzopyrylium, 3,5,7-trihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-, chloride (9CI), Flavylium, 3,4',5,7-tetrahydroxy-3',5'-dimethoxy-, chloride (8CI), Malvidin (6CI), 3,4',5,7-Tetrahydroxy-3',5'-dimethoxy-2-phenylbenzopyrylium chloride, 3,4',5,7-Tetrahydroxy-3',5'-dimethoxyflavylium chloride, Enidin, Malvidin chloride, Malvidol, Malvinidin, Malvinidol chloride, Oenidin, Primulidin, Syringidin, Syringidin chloride
IUPAC-namn 2-(4-hydroxy-3,5-dimethoxyphenyl)chromenylium-3,5,7-triol;chloride
LEDER [Cl-].COc1cc(cc(OC)c1O)c2[o+]c3cc(O)cc(O)c3cc2O
Specifikationer
Kemiskt namn eller material Malvidin Chloride
Kvantitet 5 mg
InChI formel InChI=1S/C17H14O7.ClH/c1-22-14-3-8(4-15(23-2)16(14)21)17-12(20)7-10-11(19)5-9(18)6-13(10)24-17;/h3-7H,1-2H3,(H3-,18,19,20,21);1H
Formel vikt 366.0506