Läs mer
Malvin, TRC
Produktkod. 30136806
Change view
Kvantitet:
1 mg
2.5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136806 2.5 mg 2.5mg
30012073 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136806 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1615332.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16727-30-3
Molekylformel C29 H35 O17 . Cl
Molekylvikt (g/mol) 691.03
Synonym 1-Benzopyrylium, 3,5-bis(β-D-glucopyranosyloxy)-7-hydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-, chloride (1:1), 1-Benzopyrylium, 3,5-bis(β-D-glucopyranosyloxy)-7-hydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-, chloride (9CI), Malvin (6CI,8CI), Malvidin 3,5-di-O-glucoside, Malvidin 3,5-diglucoside, Malvidol 3,5-diglucoside chloride, Malvin chloride, Malvoside, NSC 407312
IUPAC-namn (2S,3R,4S,5S,6R)-2-[7-hydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromenylium-5-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;chloride
LEDER [Cl-].COc1cc(cc(OC)c1O)c2[o+]c3cc(O)cc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c3cc2O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O
Specifikationer
Kemiskt namn eller material Malvidin-3,5-diglucoside chloride(malvin chloride)
Kvantitet 2.5 mg
InChI formel InChI=1S/C29H34O17.ClH/c1-40-15-3-10(4-16(41-2)20(15)33)27-17(44-29-26(39)24(37)22(35)19(9-31)46-29)7-12-13(42-27)5-11(32)6-14(12)43-28-25(38)23(36)21(34)18(8-30)45-28;/h3-7,18-19,21-26,28-31,34-39H,8-9H2,1-2H3,(H-,32,33);1H/t18-,19-,21-,22-,23+,24+,25-,26-,28-,29-;/m1./s1
Formel vikt 690.1563