missing translation for 'onlineSavingsMsg'
Learn More

Mammaglobin A Mouse anti-Human, Clone: 2E5D4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66237-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877925

  • 4721.67 SEK / 150µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This monoclonal antibody is raised against full-length, 93-amino acid human mammaglobin A.

Mammaglobin is a gene that is expressed almost exclusively in the normal breast epithelium and human breast cancer. It is a member of the secretoglobin gene family and forms a heterodimer with lipophilin B. It has been suggested that mammaglobin may be a useful marker for breast cancer clinical research. Studies investigating the detection of mRNA by RT PCR from circulating carcinoma cells in the peripheral blood of breast cancer patients have shown that mammaglobin is a highly specific marker and correlates with several prognostic factors, such as lymph node involvement.
Specifications

Specifications

Mammaglobin A
Monoclonal
0.87 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13296
SCGB2A2
Mammaglobin A Fusion Protein Ag22485
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
2E5D4
Unconjugated
SCGB2A2
Mammaglobin 1, Mammaglobin A, MGB1, SCGB2A2, UGB2
Mouse
Protein G
RUO
4250
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers