Learn More
CD35 Antibody (CR1/4382R), Biotin, R&D Systems™
Product Code. 18604187 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
Product Code. Quantity Unit Size
18604187 100 μg 100µL
1 options
Product Code. 18604187 Supplier Novus Biologicals Supplier No. NBP308296B
 Restricted Item
Beräknad leverans: 01-12-2026
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
CD35 Monoclonal specifically detects CD35 in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Antigen CD35
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CR1/4382R
Conjugate Biotin
Formulation PBS
Gene Alias complement component (3b/4b) receptor 1 (Knops blood group), KN
Host Species Rabbit
Immunogen Synthetic peptide corresponding to CD35 residues within aa1939-2039 of CD35 was used as an immunogen. (Uniprot: P17927 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1378
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus