Learn More
Manganese(IV) Oxide, Sigma-Aldrich™
Product Code. 15889230
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15889230 100 g 100g
15866400 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15889230 Supplier Sigma Aldrich Supplier No. 8059580100
 Restricted Item
Beräknad leverans: 19-11-2026
Precipitated active for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1313-13-9
Specifications
Quantity 100 g
Density 5.21 g/cm3 at 21°C
Molecular Formula MnO2
MDL Number MFCD00003463
Synonym Manganese(IV) oxide, Manganese dioxide, Pyrolusite, Manganese(IV) oxide
Molecular Weight (g/mol) 86.94
Grade Research
Chemical Name or Material Manganese(IV) Oxide