Learn More
Hanna Instruments™ Manganese Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Manganese Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand:  Hanna Instruments™ HI9374801
Additional Details : Weight : 0.10000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 11622660
  • 2690.00 SEK / 10mL
Estimated Shipment: 26-04-2024
to see stock.
Description
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determination of manganese. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Reagent sets available for low range (using PAN method) or high range (using Periodate method) determinations
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Specifications
1-(2-pyridylazo)-2-naphthol (PAN)
Compatible Hanna benchtop and portable photometers
50
0 to 300μg/L manganese
Water Testing Kit
Manganese low range determination
Dropper bottles and individual packets
1 Set
Cholorimetric determination