Learn More
Hanna Instruments™ Manganese Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Manganese Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand: Hanna Instruments™ HI9374801
Additional Details : Weight : 0.10000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determination of manganese. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Reagent sets available for low range (using PAN method) or high range (using Periodate method) determinations
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
1-(2-pyridylazo)-2-naphthol (PAN) | |
Compatible Hanna benchtop and portable photometers | |
50 | |
0 to 300μg/L manganese | |
Water Testing Kit |
Manganese low range determination | |
Dropper bottles and individual packets | |
1 Set | |
Cholorimetric determination |